Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs10
Entrez Gene ID
54290
Gene Name
regulator of G-protein signaling 10
Gene Aliases
-
Chromosome Location
Chr.1: 199782076 - 199823439 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019337.1 NP_062210.1 3 - 4 268 105
XM_006230325.2 XP_006230387.1 3 - 4 309 105
XM_006230326.2 XP_006230388.1 3 - 4 321 105
GenBank mRNA CD372843.1 - 3 - 4 268 105

View Full Product Details