Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adm
Entrez Gene ID
25026
Gene Name
adrenomedullin
Gene Aliases
Ap, H39316, RATAP
Chromosome Location
Chr.1: 175443208 - 175447260 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012715.1 NP_036847.1 2 - 3 268 68
XM_008759687.1 XP_008757909.1 3 - 4 1775 68
GenBank mRNA BC061775.1 - 2 - 3 264 68
D15069.1 - 2 - 3 250 68
U15419.1 - 2 - 3 268 68

View Full Product Details