The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012715.1 | NP_036847.1 | 3 - 4 | 414 | 72 |
XM_008759687.1 | XP_008757909.1 | 4 - 5 | 1921 | 72 | |
GenBank mRNA | BC061775.1 | - | 3 - 4 | 410 | 72 |
D15069.1 | - | 3 - 4 | 396 | 72 | |
U15419.1 | - | 3 - 4 | 414 | 72 |