Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnah3
Entrez Gene ID
117249
Gene Name
dynein, axonemal, heavy chain 3
Gene Aliases
Dnahc3, Dnahc3l
Chromosome Location
Chr.1: 189688826 - 189865233 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759836.1 XP_008758058.1 57 - 58 11669 98

View Full Product Details