Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Swap70
Entrez Gene ID
293410
Gene Name
SWAP switching B-cell complex 70
Gene Aliases
-
Chromosome Location
Chr.1: 174862725 - 174924100 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106288.1 NP_001099758.1 2 - 3 303 144

View Full Product Details