The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039208.2 | NP_001034297.2 | 5 - 6 | 1011 | 104 |
GenBank mRNA | BC110048.1 | - | 5 - 6 | 996 | 104 |
BU671623.1 | - | 2 - 3 | 194 | 104 | |
CA339235.1 | - | 5 - 6 | 584 | 104 |