The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039208.2 | NP_001034297.2 | 11 - 12 | 1836 | 159 |
GenBank mRNA | BC110048.1 | - | 8 - 9 | 2669 | 159 |