Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrdl2
Entrez Gene ID
308854
Gene Name
chordin-like 2
Gene Aliases
-
Chromosome Location
Chr.1: 165008997 - 165036431 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107537.1 NP_001101007.1 10 - 11 1373 87

View Full Product Details