Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim72
Entrez Gene ID
365377
Gene Name
tripartite motif-containing 72
Gene Aliases
RGD1562778
Chromosome Location
Chr.1: 199450042 - 199457902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077675.1 NP_001071143.1 4 - 5 803 93
GenBank mRNA BC127539.1 - 4 - 5 803 93
FQ224850.1 - 4 - 5 844 93

View Full Product Details