Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coro1a
Entrez Gene ID
155151
Gene Name
coronin, actin binding protein 1A
Gene Aliases
-
Chromosome Location
Chr.1: 198123883 - 198128857 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130411.2 NP_569095.1 8 - 9 1110 65
XM_006230204.2 XP_006230266.1 8 - 9 1194 65
GenBank mRNA AF416730.1 - 7 - 8 1006 65
AF495469.1 - 8 - 9 1110 65
BC086971.1 - 8 - 9 1086 65
FQ225360.1 - 5 - 6 632 65
FQ225522.1 - 4 - 5 549 65
FQ226379.1 - 8 - 9 1101 65
FQ226941.1 - 8 - 9 1130 65
FQ233306.1 - 8 - 9 1104 65
FQ233390.1 - 8 - 9 1106 65
FQ233980.1 - 8 - 9 1024 65
FQ234750.1 - 8 - 9 1112 65

View Full Product Details