Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
301221
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
-
Chromosome Location
Chr.9: 13493705 - 13520113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106881.1 NP_001100351.1 6 - 7 836 64
XM_006244415.2 XP_006244477.1 7 - 8 906 64
XM_006244416.2 XP_006244478.1 6 - 7 847 64
XM_006244418.2 XP_006244480.1 7 - 8 901 64
XM_008766840.1 XP_008765062.1 3 - 4 254 64
XM_008766841.1 XP_008765063.1 6 - 7 845 64

View Full Product Details