The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031547.1 | NP_113735.1 | 2 - 2 | 2622 | 105 |
XM_008762022.1 | XP_008760244.1 | 2 - 2 | 2524 | 105 | |
GenBank mRNA | CR472483.1 | - | 1 - 1 | 105 | |
M16409.1 | - | 1 - 1 | 2443 | 105 |