Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scn2a
Entrez Gene ID
24766
Gene Name
sodium channel, voltage-gated, type II, alpha subunit
Gene Aliases
NachII, Nav1.2, RII/RIIA, RNSCPIIR, SCN, Scn2a1, Scn2a2, ScpII
Chromosome Location
Chr.3: 51687910 - 51822008 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012647.1 NP_036779.1 21 - 22 4177 62
GenBank mRNA M22254.1 - 21 - 22 4176 62
X03639.1 - 21 - 22 4177 62

View Full Product Details