Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp1r1c
Entrez Gene ID
499818
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 1C
Gene Aliases
RGD1562095
Chromosome Location
Chr.3: 66673071 - 66775671 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109200.1 NP_001102670.1 2 - 3 540 68

View Full Product Details