Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tank
Entrez Gene ID
252961
Gene Name
TRAF family member-associated NFKB activator
Gene Aliases
-
Chromosome Location
Chr.3: 47439006 - 47515706 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164073.1 NP_001157545.1 1 - 2 81 86
XM_006234273.2 XP_006234335.1 1 - 2 151 86
GenBank mRNA BC129086.1 - 1 - 2 81 86

View Full Product Details