The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077429.1 | NP_001070897.1 | 13 - 14 | 1237 | 121 |
XM_006234385.2 | XP_006234447.1 | 13 - 14 | 1321 | 121 | |
GenBank mRNA | BC126100.1 | - | 13 - 14 | 1237 | 121 |