The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111294.1 | NP_001104764.1 | 5 - 6 | 724 | 66 |
NM_001111295.1 | NP_001104765.1 | 5 - 6 | 658 | 66 | |
NM_198132.3 | NP_937765.1 | 5 - 6 | 724 | 66 | |
XM_008761911.1 | XP_008760133.1 | 5 - 6 | 673 | 66 | |
XM_008761912.1 | XP_008760134.1 | 5 - 6 | 673 | 66 | |
XM_008761913.1 | XP_008760135.1 | 5 - 6 | 719 | 66 | |
GenBank mRNA | AY363226.1 | - | 5 - 6 | 670 | 66 |
AY363227.1 | - | 5 - 6 | 604 | 66 | |
BC081878.1 | - | 5 - 6 | 664 | 66 | |
CB725005.1 | - | 5 - 6 | 596 | 66 | |
FQ214617.1 | - | 5 - 6 | 620 | 66 | |
Y16641.1 | - | 1 - 2 | 86 | 66 |