The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108699.1 | NP_001102169.1 | 5 - 6 | 933 | 100 |
GenBank mRNA | BC079468.1 | - | 5 - 6 | 1559 | 100 |
FQ220869.1 | - | 4 - 5 | 405 | 100 |