The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107978.2 | NP_001101448.1 | 7 - 8 | 746 | 103 |
GenBank mRNA | BC168964.1 | - | 7 - 8 | 746 | 103 |
FQ226285.1 | - | 7 - 8 | 758 | 103 | |
FQ231750.1 | - | 7 - 8 | 767 | 103 | |
FQ233930.1 | - | 7 - 8 | 758 | 103 |