Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tas1r1
Entrez Gene ID
29407
Gene Name
taste receptor, type 1, member 1
Gene Aliases
Gpr70
Chromosome Location
Chr.5: 169200389 - 169212113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053305.1 NP_445757.1 3 - 4 1258 101
XM_008764341.1 XP_008762563.1 3 - 4 1118 101
GenBank mRNA AF127389.1 - 3 - 4 1258 101

View Full Product Details