Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb17
Entrez Gene ID
313666
Gene Name
zinc finger and BTB domain containing 17
Gene Aliases
Zfp100
Chromosome Location
Chr.5: 159993799 - 160014899 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012105.1 NP_001012105.1 5 - 6 814 85
XM_006239303.1 XP_006239365.1 5 - 6 788 85
XM_008764253.1 XP_008762475.1 4 - 5 699 85
GenBank mRNA BC083577.1 - 5 - 6 814 85

View Full Product Details