Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rbbp4
- Entrez Gene ID
-
313048
- Gene Name
- retinoblastoma binding protein 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 147499880 - 147535240 on Build Rnor_6.0
- Gene Symbol
- RGD1563620
- Entrez Gene ID
-
310511
- Gene Name
- similar to retinoblastoma binding protein 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 166348373 - 166350275 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_227252.8 |
XP_227252.5 |
3 - 3 |
1221 |
113 |
View Full Product Details