Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfyve1
Entrez Gene ID
299188
Gene Name
zinc finger, FYVE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 107031411 - 107080587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106743.1 NP_001100213.1 10 - 11 2169 65
XM_006240323.2 XP_006240385.1 10 - 11 2643 65
XM_006240324.2 XP_006240386.1 10 - 11 2639 65
XM_006240325.2 XP_006240387.1 10 - 11 2584 65
XM_008764752.1 XP_008762974.1 7 - 8 1556 65
XM_008764753.1 XP_008762975.1 7 - 8 1732 65

View Full Product Details