Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfyve1
Entrez Gene ID
299188
Gene Name
zinc finger, FYVE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 107031411 - 107080587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106743.1 NP_001100213.1 4 - 5 1377 98
XM_006240323.2 XP_006240385.1 4 - 5 1851 98
XM_006240324.2 XP_006240386.1 4 - 5 1847 98
XM_006240325.2 XP_006240387.1 4 - 5 1792 98

View Full Product Details