Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnal1
Entrez Gene ID
685664
Gene Name
dynein, axonemal, light chain 1
Gene Aliases
Dnalc1
Chromosome Location
Chr.6: 107596785 - 107623869 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240352.2 XP_006240414.1 2 - 3 257 102

View Full Product Details