Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxn3
Entrez Gene ID
314374
Gene Name
forkhead box N3
Gene Aliases
Ches1
Chromosome Location
Chr.6: 123183720 - 123577695 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108047.1 NP_001101517.1 1 - 2 110 74

View Full Product Details