Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxn3
Entrez Gene ID
314374
Gene Name
forkhead box N3
Gene Aliases
Ches1
Chromosome Location
Chr.6: 123183720 - 123577695 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108047.1 NP_001101517.1 5 - 6 936 76
XM_008764803.1 XP_008763025.1 4 - 5 2309 76

View Full Product Details