The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006233151.2 | XP_006233213.1 | 3 - 4 | 283 | 83 |
GenBank mRNA | BC166866.1 | - | 3 - 4 | 247 | 83 |
FM135814.1 | - | 3 - 4 | 221 | 83 |