The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053757.1 | NP_446209.1 | 5 - 6 | 664 | 92 |
GenBank mRNA | AF320763.1 | - | 1 - 2 | 88 | 92 |
BC098675.1 | - | 5 - 6 | 664 | 92 | |
FQ226969.1 | - | 3 - 4 | 440 | 92 | |
FQ229959.1 | - | 3 - 4 | 440 | 92 |