Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ldlrap1
Entrez Gene ID
500564
Gene Name
low density lipoprotein receptor adaptor protein 1
Gene Aliases
RGD1563417
Chromosome Location
Chr.5: 152964294 - 152987286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109271.1 NP_001102741.1 2 - 3 305 65

View Full Product Details