The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025773.1 | NP_001020944.1 | 7 - 8 | 1100 | 69 |
XM_006239471.2 | XP_006239533.1 | 8 - 9 | 1520 | 69 | |
XM_006239472.2 | XP_006239534.1 | 7 - 8 | 1953 | 69 | |
XM_008764282.1 | XP_008762504.1 | 7 - 8 | 1290 | 69 | |
XM_008764283.1 | XP_008762505.1 | 8 - 9 | 1541 | 69 | |
GenBank mRNA | AB164643.1 | - | 6 - 7 | 676 | 69 |
BC097483.1 | - | 7 - 8 | 1100 | 69 |