Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apitd1
Entrez Gene ID
100534597
Gene Name
apoptosis-inducing, TAF9-like domain 1
Gene Aliases
-
Chromosome Location
Chr.5: 165946106 - 165955723 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204879.1 NP_001191808.1 4 - 5 299 77
GenBank mRNA BI282680.1 - 1 - 2 77
FM092845.1 - 4 - 5 298 77

View Full Product Details