Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Epha10
Entrez Gene ID
298528
Gene Name
EPH receptor A10
Gene Aliases
RGD1562679
Chromosome Location
Chr.5: 142875683 - 142910537 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135709.1 NP_001129181.1 13 - 14 2239 69
XM_006238858.2 XP_006238920.1 11 - 12 2231 69

View Full Product Details