Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thap3
Entrez Gene ID
362667
Gene Name
THAP domain containing, apoptosis associated protein 3
Gene Aliases
RGD1308109
Chromosome Location
Chr.5: 169155239 - 169160987 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108695.1 NP_001102165.1 3 - 4 487 72
GenBank mRNA BC159427.1 - 3 - 4 385 72
FQ229895.1 - 3 - 4 373 72

View Full Product Details