Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptchd2
Entrez Gene ID
313705
Gene Name
patched domain containing 2
Gene Aliases
RGD1305820
Chromosome Location
Chr.5: 165051522 - 165099823 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750087.3 XP_003750135.1 16 - 17 3504 80

View Full Product Details