Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Extl1
Entrez Gene ID
313610
Gene Name
exostosin-like glycosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.5: 152573755 - 152588956 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107985.1 NP_001101455.1 1 - 2 831 118

View Full Product Details