Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Depdc1
Entrez Gene ID
295538
Gene Name
DEP domain containing 1
Gene Aliases
Depdc1a
Chromosome Location
Chr.2: 264704734 - 264738265 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233537.2 XP_006233599.1 6 - 7 884 151
XM_227798.8 XP_227798.5 6 - 7 886 151

View Full Product Details