The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025018.1 | NP_001020189.1 | 8 - 9 | 947 | 93 |
XM_006233120.2 | XP_006233182.1 | 8 - 9 | 891 | 93 | |
XM_006233121.2 | XP_006233183.1 | 7 - 8 | 831 | 93 | |
XM_006233122.2 | XP_006233184.1 | 7 - 8 | 932 | 93 | |
GenBank mRNA | BC091251.1 | - | 8 - 9 | 947 | 93 |
FQ218349.1 | - | 8 - 9 | 959 | 93 | |
FQ228782.1 | - | 8 - 9 | 1044 | 93 |