Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcoln2
Entrez Gene ID
292168
Gene Name
mucolipin 2
Gene Aliases
-
Chromosome Location
Chr.2: 252018594 - 252068209 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039005.1 NP_001034094.1 4 - 5 620 112
GenBank mRNA BC099787.1 - 4 - 5 620 112

View Full Product Details