Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lef1
Entrez Gene ID
161452
Gene Name
lymphoid enhancer binding factor 1
Gene Aliases
-
Chromosome Location
Chr.2: 236232117 - 236345061 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233305.2 XP_006233367.1 5 - 6 1951 93
XM_006233306.2 XP_006233368.1 5 - 6 1950 93
XM_008761490.1 XP_008759712.1 5 - 6 1953 93
GenBank mRNA EF519319.1 - 2 - 3 121 93

View Full Product Details