Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csf1
Entrez Gene ID
78965
Gene Name
colony stimulating factor 1 (macrophage)
Gene Aliases
-
Chromosome Location
Chr.2: 210522370 - 210550546 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023981.4 NP_076471.3 6 - 7 1760 109
XM_006233187.1 XP_006233249.1 7 - 8 1712 109
GenBank mRNA AF514355.1 - 6 - 7 1600 109
AF514356.1 - 6 - 7 1600 109
AF514357.1 - 6 - 7 1610 109
AF515736.1 - 6 - 7 1698 109
BC074007.1 - 6 - 7 1760 109

View Full Product Details