Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csf1
Entrez Gene ID
78965
Gene Name
colony stimulating factor 1 (macrophage)
Gene Aliases
-
Chromosome Location
Chr.2: 210522370 - 210550546 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233187.1 XP_006233249.1 9 - 10 1828 69
XM_008761429.1 XP_008759651.1 9 - 10 1881 69

View Full Product Details