Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pigk
Entrez Gene ID
295543
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class K
Gene Aliases
-
Chromosome Location
Chr.2: 257911100 - 257997735 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011953.1 NP_001011953.1 6 - 7 592 72
XM_008761549.1 XP_008759771.1 6 - 7 618 72
GenBank mRNA BC083636.1 - 6 - 7 592 72
FQ219584.1 - 1 - 2 69 72

View Full Product Details