Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npsr1
Entrez Gene ID
300458
Gene Name
neuropeptide S receptor 1
Gene Aliases
RGD1564154
Chromosome Location
Chr.8: 25246174 - 25483582 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106808.1 NP_001100278.1 4 - 5 466 93
XM_008766049.1 XP_008764271.1 3 - 4 709 93

View Full Product Details