The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006242777.2 | XP_006242839.1 | 7 - 8 | 1062 | 60 |
XM_006242778.2 | XP_006242840.1 | 2 - 3 | 3633 | 60 | |
XM_008766070.1 | XP_008764292.1 | 11 - 12 | 1553 | 60 | |
XM_008766071.1 | XP_008764293.1 | 11 - 12 | 1553 | 60 | |
XM_008766072.1 | XP_008764294.1 | 10 - 11 | 1511 | 60 | |
XM_008766073.1 | XP_008764295.1 | 10 - 11 | 1511 | 60 | |
XM_008766074.1 | XP_008764296.1 | 11 - 12 | 2637 | 60 |