The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012630.2 | NP_036762.1 | 9 - 10 | 1041 | 105 |
GenBank mRNA | BC092133.1 | - | 9 - 10 | 1035 | 105 |
M19304.1 | - | 9 - 10 | 1042 | 105 |