The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109025.2 | NP_001102495.1 | 1 - 2 | 242 | 114 |
GenBank mRNA | BC088341.1 | - | 1 - 2 | 218 | 114 |
DY573537.1 | - | 1 - 2 | 242 | 114 |