Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfb
Entrez Gene ID
294257
Gene Name
complement factor B
Gene Aliases
Bf, Da1-24
Chromosome Location
Chr.20: 4536206 - 4542073 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212466.3 NP_997631.2 8 - 9 1376 85
GenBank mRNA BC087089.1 - 8 - 9 1376 85

View Full Product Details