Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmr1nb
Entrez Gene ID
293918
Gene Name
fragile X mental retardation 1 neighbor
Gene Aliases
RGD1564059
Chromosome Location
Chr.X: 154631546 - 154653776 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229540.2 XP_006229602.1 4 - 5 706 165
XM_215239.7 XP_215239.2 5 - 6 724 165

View Full Product Details