Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccny
Entrez Gene ID
361261
Gene Name
cyclin Y
Gene Aliases
RGD1565969
Chromosome Location
Chr.17: 62332537 - 62462644 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191833.1 NP_001178762.1 9 - 10 911 58

View Full Product Details